Merck
  • Cholesterol-based polymeric monolithic columns for capillary liquid chromatography.
Cholesterol-based polymeric monolithic columns for capillary liquid chromatography.
Journal of chromatography. A (2014-12-04)
Michał Szumski, Damian Grzywiński, Bogusław Buszewski
ABSTRACT
A novel, cholesterol-based polymeric monolithic stationary phase for capillary liquid chromatography, was prepared by thermally initiated in-situ polymerization. Cholesteryl methacrylate (CholMA) was used as a functional monomer and trimethylolpropane trimethacrylate (TRIM) was a cross-linker, while azobisisobutyronitrile (AIBN) was an initiator. Isooctane and toluene were chosen as "poor" and "good" solvent, respectively, as constituents of the porogen solvent. Isocratic elutions of alkylbenzenes and separation of the testing mixture of o-terphenyl and triphenylene were conducted for all of the monoliths to assess their hydrophobicity and planar selectivity characteristic for cholesterol-based stationary phases. The synthesized columns demonstrated efficiency exceeding N=10,000 plates and a plate height of ca. H=30 μm. Column preparation was found to be highly reproducible; the relative standard deviation (RSD) values (n=3) for day-to-day and column-to-column were less than 4.08 and 2.02%, respectively, based on retention factor of alkylbenzenes.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
o-Terphenyl, 99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Triethylamine, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Cyclohexane, analytical standard