Merck
  • Lipophilic prodrugs of apomorphine I: preparation, characterisation, and in vitro enzymatic hydrolysis in biorelevant media.
Lipophilic prodrugs of apomorphine I: preparation, characterisation, and in vitro enzymatic hydrolysis in biorelevant media.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-17)
Nrupa Borkar, Boyang Li, René Holm, Anders E Håkansson, Anette Müllertz, Mingshi Yang, Huiling Mu
ABSTRACT
Apomorphine, a subcutaneously administered drug for Parkinson's disease with short half-life requires frequent administration leading to patient non-compliance. This study aimed at synthesising and purifying lipophilic diesters of apomorphine, and investigating their in vitro degradation in biorelevant media before and after incorporating them into self-emulsifying drug delivery systems (SEDDS) for oral delivery. Two apomorphine diester prodrugs were synthesised: dilauroyl apomorphine (DLA) and dipalmitoyl apomorphine (DPA). The in vitro enzymatic hydrolysis of diesters was performed using biorelevant media with pancreatin to catalyse the diester degradation. The synthesised and purified diesters were found to be free from reactants as impurities confirmed by LC/MS and NMR. DLA and DPA were degraded into corresponding monoesters and free apomorphine within 5 min after adding pancreatin, leaving about 4% and 28% of the intact diester, respectively. The incorporation of the diesters into SEDDS reduced the enzymatic degradation of diesters. In addition, the chain length of diester and the type of oil used in formulations affected diester hydrolysis. The lipophilic apomorphine diesters were substrates of lipases present in pancreatin, and the degree of diester degradation can be controlled by selecting suitable lipid excipients. Therefore, diesters of apomorphine are promising prodrugs for oral delivery aiming at lymphatic transport.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Palmitoyl chloride, 98%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)