Merck
von Willebrand disease and aging: an evolving phenotype.
Journal of thrombosis and haemostasis : JTH (2014-04-23)
Y V Sanders, M A Giezenaar, B A P Laros-van Gorkom, K Meijer, J G van der Bom, M H Cnossen, M R Nijziel, P F Ypma, K Fijnvandraat, J Eikenboom, E P Mauser-Bunschoten, F W G Leebeek
ABSTRACT
Because the number of elderly von Willebrand disease (VWD) patients is increasing, the pathophysiology of aging in VWD has become increasingly relevant. To assess age-related changes in von Willebrand factor (VWF) and factor VIII (FVIII) levels and to compare age-related differences in bleeding phenotype between elderly VWD patients and those < 65 years. We also studied co-morbidity in elderly patients. We included VWD patients with VWF levels ≤ 30 U dL(-1) in the nationwide cross-sectional 'Willebrand in the Netherlands' (WiN-) study. Patients reported bleeding episodes and treatment of VWD in the year preceding inclusion and during life. This was compared between VWD patients older (n = 71) and younger (16-64 years, n = 593) than 65 years. In elderly patients, age-related changes in VWF and FVIII levels were studied longitudinally by including all historically measured levels. All medical records were examined for co-morbidity. In elderly type 1 patients, a decade age increase was associated with a 3.5 U dL(-1) (95% CI, -0.6 to 7.6) VWF:Ag increase and 7.1 U dL(-1) (95% CI, 0.7 to 13.4) FVIII:C increase. This increase was not observed in elderly type 2 patients. Elderly type 2 patients reported significantly more bleeding symptoms in the year preceding inclusion than younger patients (16/27, 59% vs. 87/221, 39%; P = 0.048), which was not observed in type 1 VWD. von Willebrand factor parameters and bleeding phenotype evolve with increasing age in VWD. VWF and FVIII levels increase with age in type 1 patients with no mitigation in bleeding phenotype. In type 2 patients VWF parameters do not increase with age and in these patients aging is accompanied by increased bleeding.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material