Merck
  • Rapid quantitative determination of related substances and degradants in milnacipran.
Rapid quantitative determination of related substances and degradants in milnacipran.
Journal of chromatographic science (2012-12-22)
Surya Prakash Rao Pydimarry, Vijay Kumar Cholleti, Ranga Reddy Venagala
ABSTRACT
A simple, sensitive, precise and accurate stability-indicating ultra-performance liquid chromatography (UPLC) method was developed for the quantitative determination of the purity of the active pharmaceutical ingredient of milnacipran hydrochloride (MCP) and its pharmaceutical dosage form in the presence of its process-related impurities and degradation products. The proposed UPLC method utilizes Acquity UPLC BEH 100 mm, 2.1 mm and 1.7 µm phenyl columns at 27°C with a gradient program of 12.0 min at a flow rate of 0.5 mL/min. The compounds of interest were monitored at 210 nm. The resolution for MCP and 12 (potential byproducts and degradation) impurities was found to be greater than 2.0 for any pair of components. The high correlation coefficients (r(2) >0.9995) indicate clear correlations between the concentrations of investigated compounds and their peak areas. The relative standard deviations obtained for the repeatability and intermediate precision experiments were less than 5.0%. The accuracy of the method was further ascertained by performing recovery studies through spiking experiments. The drug substance was subjected to hydrolytic, oxidative, photolytic and thermal stress conditions, as per the International Conference on Harmonization (ICH). The developed method was validated according to the current ICH guidelines for specificity, limits of detection and quantitation, linearity, accuracy, precision, ruggedness and robustness. The method is also suitable for the assay determination of MCP in pharmaceutical dosage forms.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)