Merck
Improved delivery of the natural anticancer drug tetrandrine.
International journal of pharmaceutics (2014-12-17)
Chen Shi, Saeed Ahmad Khan, Kaiping Wang, Marc Schneider
ABSTRACT
The study aims at designing a nanoparticle-based delivery system to improve the efficacy of the natural compound tetrandrine against lung cancer. Nanoparticles from poly(lactic-co-glycolic acid) (PLGA) were prepared by the emulsion solvent diffusion method and characterized for their physicochemical properties and drug-loading efficiency. Furthermore, the cellular uptake and the anti-cancerous activity was studied on A549 cell line. To investigate the surface properties and uptake, three different stabilizers were used to analyze the effect on size and zeta potential of nanoparticles as well as the effect on the cellular uptake. Nanoparticles in the size range of 180-200 nm with spherical shape were obtained with polyvinyl alcohol (PVA), Pluronic-F127 (PF127) and didodecyldimethylammonium bromide (DMAB), 2%, 1% and 0.1%, respectively. An entrapment efficiency of 50-60% with a loading of 1.5-2% was observed. In vitro release profile at pH 7.4 PBS solution showed a consistent release over 168 h. All particle systems showed an improved performance over the pure drug at the same drug concentration. DMAB stabilized particles demonstrated the most pronounced effect against A549 cells compared to pure drug while PVA stabilized particles were least effective in terms of antitumor activity.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)