Merck
  • Reliability of computer-assisted method transfer between several column dimensions packed with 1.3-5μm core-shell particles and between various instruments.
Reliability of computer-assisted method transfer between several column dimensions packed with 1.3-5μm core-shell particles and between various instruments.
Journal of pharmaceutical and biomedical analysis (2014-03-08)
Róbert Kormány, Jenő Fekete, Davy Guillarme, Szabolcs Fekete
ABSTRACT
In this contribution, the possibility to automatically transfer RPLC methods between different column dimensions and instruments was evaluated using commercial modelling software. The method transfer reliability was tested with loratadine and its 7 related pharmacopeial impurities. In this study, state-of-the-art columns packed with superficially porous particles of 5, 2.6, 1.7 and 1.3μm particles were exclusively employed. A fast baseline separation of loratadine and related impurities (Rs,min=2.49) was achieved under the best analytical conditions (i.e. column of 50mm×2.1mm, 1.3μm, 10-90% ACN in 5min, T=40°C, pH=3, F=0.5ml/min). This optimal method was successfully tested on columns packed with other particle sizes, namely 1.7 and 2.6μm, to reduce pressure drop. The selectivities and retentions remained identical, while the peak widths were logically wider, leading to a reduction of peak capacity from 203 to 181 and 159 on the 1.3, 1.7 and 2.6μm particles, respectively. On the minimum, the resolution was equal to 1.54 on the 50mm×2.1mm, 2.6μm stationary phase. Next to this, the method was transferred to columns of different lengths, inner diameters and particle sizes (100mm×3mm, 2.6μm or 150mm×4.6mm, 5μm). These columns were used on other LC instruments possessing larger dwell volumes. The modelling software employed for developing the original method was able to calculate the new gradient conditions to be used. The accuracy of prediction was excellent, as the average retention time errors between predicted and observed chromatograms were -0.11% and 0.45% when transferring the method to 100mm×3mm and 150mm×4.6mm columns, respectively. This work proves the usefulness and validity of HPLC modelling software for transferring methods between different instruments, column dimensions and/or flow rates.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Loratadine, ≥98% (HPLC), powder
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
USP
Loratadine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Loratadine, Pharmaceutical Secondary Standard; Certified Reference Material
Loratadine, European Pharmacopoeia (EP) Reference Standard
Loratadine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)