Merck
  • Anti-proliferative activity of nano-formulated phenolato titanium(IV) complexes against cancer cells.
Anti-proliferative activity of nano-formulated phenolato titanium(IV) complexes against cancer cells.
ChemMedChem (2014-03-29)
Sigalit Meker, Katrin Margulis-Goshen, Ester Weiss, Ori Braitbard, Jacob Hochman, Shlomo Magdassi, Edit Y Tshuva
ABSTRACT
Nanoparticles of titanium(IV) complexes of phenolato ligands were formed and evaluated for cytotoxicity toward human HT-29 colon cancer, murine T-25 lymphoma, and murine HU-2 multidrug-resistant (MDR) cells. The nano-formulation, besides increasing the complexes' shelf lives, is particularly efficient in overcoming limitations in solubility and cell-penetration, thus enhancing biological accessibility; large complexes that were inactive when measured in a non-formulated form showed marked activity when nano-formulated. For active and accessible small complexes, the effect of the formulation was negligible. Most complexes showed similar activity toward MDR cells and their drug-sensitive analogues, further increasing their therapeutic potential. An exception is a particularly hydrophobic complex, which is presumably more accessible to interaction with the membrane ABCB1 (MDR1) transporter active in the multidrug resistance of HU-2 cells. The most efficient compound is a mononuclear complex of a single hexadentate ligand, combining particularly high activity and hydrolytic stability with accessibility aided by the nano-formulation.
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)