Merck
  • The visualization of large organized chromatin domains enriched in the H3K9me2 mark within a single chromosome in a single cell.
The visualization of large organized chromatin domains enriched in the H3K9me2 mark within a single chromosome in a single cell.
Epigenetics (2014-12-09)
Xingqi Chen, Samer Yammine, Chengxi Shi, Mariliis Tark-Dame, Anita Göndör, Rolf Ohlsson
ABSTRACT
Despite considerable efforts, our understanding of the organization of higher order chromatin conformations in single cells and how these relate to chromatin marks remains poor. We have earlier invented the Chromatin In Situ Proximity (ChrISP) technique to determine proximities between chromatin fibers within a single chromosome. Here we used ChrISP to identify chromosome 11-specific hubs that are enriched in the H3K9me2 mark and that project toward the nuclear membrane in finger-like structures. Conversely, chromosome 11-specfic chromatin hubs, visualized by the presence of either H3K9me1 or H3K9me3 marks, are chromosome-wide and largely absent at the nuclear periphery. As the nuclear periphery-specific chromatin hubs were lost in the induced reduction of H3K9me2 levels, they likely represent Large Organization Chromatin in Lysine Methylation (LOCK) domains, previously identified by ChIP-seq analysis. Strikingly, the downregulation of the H3K9me2/3 marks also led to the chromosome-wide compaction of chromosome 11, suggesting a pleiotropic function of these features not recognized before. The ChrISP-mediated visualization of dynamic chromatin states in single cells thus provides an analysis of chromatin structures with a resolution far exceeding that of any other light microscopic technique.
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, for molecular biology
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Roche
Digoxigenin-11-dUTP, alkali-labile, =85% (HPLC), solution, pkg of 25 μL (11573152910 [1 mM]), pkg of 125 μL (11573179910 [1 mM]]])
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®