Merck
  • Multifaceted interaction of the traditional Chinese medicinal herb Schisandra chinensis with cytochrome P450-mediated drug metabolism in rats.
Multifaceted interaction of the traditional Chinese medicinal herb Schisandra chinensis with cytochrome P450-mediated drug metabolism in rats.
Journal of ethnopharmacology (2014-08-06)
Baolian Wang, Shuang Yang, Jinping Hu, Yan Li
ABSTRACT
Schisandra chinensis (SC), officially listed as a sedative and tonic in the Chinese Pharmacopoeia, has been used as a common component in various prescriptions in Traditional Chinese Medicine (TCM) and more recently in western medicine for its antihepatotoxic effect. To assess the possible herb-drug interaction, effects of SC extracts on hepatic cytochrome P450 (P450, CYP) enzymes were studied. Effects of SC extracts on rat hepatic CYP450 enzymes in vitro and in vivo were investigated by probe substrates method, real-time RT-PCR assay and Western blotting analysis. Furthermore, the effects of SC alcoholic extract on the PK of four SC lignans and the drugs possibly co-administrated in vivo were studied in male Sprague-Dawley rat. SC aqueous extract and alcoholic extract showed significant inhibitory effect on the activities of rat liver microsomal CYP1A2, 2C6, 2C11, 2D2, 2E1 and 3A1/2 in vitro. Multiple administrations of SC aqueous extract (1.5g/kg, qd×7d) and alcoholic extract (1.5g/kg, qd×7d) increased the activities, mRNA and protein expressions of CYP2E1 and CYP3A1/2, and meanwhile, inhibited the activities and mRNA expression of CYP2D2 in vivo. The in vivo metabolism of four SC lignans, such as schisandrin, schisantherin A, deoxyshisandrin and γ-schisandrin, and chlorzoxazone was significantly accelerated, exhibited by the reduced AUC and increased CLz/F, by 7-day pretreatment with SC alcoholic extract. However, both single and multiple dosing treatments of SC alcoholic extract remarkably decreased the in vivo metabolism of tacrolimus indicated by the enhanced AUC (7-12 fold) and elevated Cmax (10 fold). These results revealed that the SC extracts exhibited multifaceted effects on rat hepatic CYP450 enzymes. Herb-drug interaction should be paid intense attention between SC components and drugs metabolized by different CYP450 enzymes.
Product Description
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
Acetaminophen, analytical standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material