Merck
  • Cyclosporine A enhances gluconeogenesis while sirolimus impairs insulin signaling in peripheral tissues after 3 weeks of treatment.
Cyclosporine A enhances gluconeogenesis while sirolimus impairs insulin signaling in peripheral tissues after 3 weeks of treatment.
Biochemical pharmacology (2014-06-25)
P C Lopes, A Fuhrmann, F Carvalho, J Sereno, M R Santos, M J Pereira, J W Eriksson, F Reis, E Carvalho
ABSTRACT
Cyclosporine A (CsA) and sirolimus (SRL) are immunosuppressive agents (IA) associated with new-onset diabetes after transplantation (NODAT). This study aims to evaluate the effects of 3-weeks of treatment with either CsA (5 mg/kg BW/day) or SRL (1 mg/kg BW/day) on insulin signaling and expression of markers involved in glucose metabolism in insulin-sensitive tissues, in Wistar rats. Although no differences were observed in fasting glucose, insulin or C-peptide levels, both treated groups displayed an impaired glucose excursion during both glucose and insulin tolerance tests. These results suggest glucose intolerance and insulin resistance. An increase in glucose-6-phosphatase protein levels (68%, p < 0.05) and in protein-tyrosine phosphatase 1B (163%, p < 0.05), a negative regulator of insulin was observed in the CsA-treated group in the liver, indicating enhanced gluconeogenesis and increased insulin resistance. On the other hand, glucokinase protein levels were decreased in the SRL group (35%, p < 0.05) compared to vehicle, suggesting a decrease in glucose disposal. SRL treatment also reduced peroxisome proliferator-activated receptor γ coactivator 1 alpha protein expression in muscle (~50%, p < 0.05), while no further protein alterations were observed in muscle and perirenal adipose tissue nor with the CsA treatment. Moreover, the phosphorylation of key proteins of the insulin signaling cascade was suppressed in the SRL group, but was unchanged by the CsA treatment. Taken together, these data suggest that CsA treatment enhances gluconeogenic factors in liver, while SRL treatment impairs insulin signaling in peripheral tissues, which can contribute to the development of insulin resistance and NODAT associated with immunosuppressive therapy.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Methanol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer