Merck
  • Development of a validated stability-indicating HPTLC method for rufinamide in bulk and its pharmaceutical dosage form.
Development of a validated stability-indicating HPTLC method for rufinamide in bulk and its pharmaceutical dosage form.
Journal of chromatographic science (2013-12-04)
Apexa Patel, Arpit Patwari, Bhanubhai Suhagia
ABSTRACT
A sensitive, selective, precise and stability indicating a high-performance thin layer chromatographic method for the analysis of rufinamide (Rf) in bulk drug and its formulations was developed and validated. The method employed thin layer chromatography aluminum plates precoated with silica gel 60 F254 as the stationary phase. The solvent system consisted of chloroform : methanol : glacial acetic acid (9 : 1 : 0.1 v/v/v). This system was found to give compact spots for Rf (Rf value of 0.68 ± 0.02). Rf was subjected to acid and alkali hydrolysis, oxidation, photodegradation and dry heat treatment. Also, the degraded products were well separated from the pure drug. Densitometric analysis of Rf was carried out in the absorbance mode at 210 nm, which is wavelength maxima for the degradant. The linear regression data for the calibration plots showed a good linear relationship with an r(2) of 0.9989 in the concentration range of 1,000-3,500 ng. The method was validated for precision, accuracy, ruggedness and recovery. The limits of detection and quantitation were 196.59 and 595.74 ng spot(-1), respectively. The drug undergoes degradation under acidic and basic conditions. All the peaks of degraded products were resolved from the standard drug with significantly different Rf values.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%