Merck
  • Simultaneous tissue profiling of eicosanoid and endocannabinoid lipid families in a rat model of osteoarthritis.
Simultaneous tissue profiling of eicosanoid and endocannabinoid lipid families in a rat model of osteoarthritis.
Journal of lipid research (2014-07-27)
Amy Wong, Devi R Sagar, Catharine A Ortori, David A Kendall, Victoria Chapman, David A Barrett
ABSTRACT
We describe a novel LC method for the simultaneous and quantitative profiling of 43 oxylipins including eicosanoids, endocannabinoids, and structurally related bioactive lipids with modified acyl groups. The LC-MS/MS method uses switching at a defined time between negative and positive electrospray ionization modes to achieve optimal detection sensitivity for all the lipids. The validated method is linear over a range of 0.01-5 nmol/g (0.1-50 nmol/g for 2-arachidonoyl glycerol) with intra- and interday precision and accuracy between 1.38 and 26.76% and 85.22 and 114.3%, respectively. The method successfully quantified bioactive lipids in different tissue types in the rat, including spinal cord, dorsal root ganglia (DRGs), knee joint, brain, and plasma. Distinct regional differences in the pattern of lipid measured between tissue types were observed using principle component analysis. The method was applied to analyze tissue samples from an established preclinical rat model of osteoarthritis (OA) pain and showed that levels of 12-hydroxyeicosatetraenoic acid were significantly increased in the OA rat knee joint compared with controls, and that 15-hydroxyeicosatetraenoic acid was significantly increased in the DRGs in the model of OA compared with controls. The developed LC-MS/MS method has the potential to provide detailed pathway profiling in tissues and biofluids where the disruption of bioactive oxylipins may be involved in disease states.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Ethyl acetate, analytical standard