Merck
  • GC-MS and FTIR evaluation of the six benzoyl-substituted-1-pentylindoles: isomeric synthetic cannabinoids.
GC-MS and FTIR evaluation of the six benzoyl-substituted-1-pentylindoles: isomeric synthetic cannabinoids.
Talanta (2014-08-17)
Forrest T Smith, Jack DeRuiter, Karim Abdel-Hay, C Randall Clark
ABSTRACT
This report compares the GC-MS and FTIR properties of all 6 regioisomeric benzoyl substituted-1-n-pentylindoles. These compounds have the benzoyl-group attached at each of the possible ring substituent positions of the indole ring. The six compounds have the same elemental composition C20H21NO yielding identical nominal and exact masses. Additionally, the substituents attached to the indole ring, benzoyl- and 1-n-pentyl-groups, are identical for all six isomers. The electron ionization mass spectra show equivalent regioisomeric major fragments resulting from cleavage of the groups attached to the central indole nucleus. Fragment ions occur at m/z 77 and 105 for the phenyl and benzoyl cations common to all six regioisomeric substances. Fragmentation of the benzoyl and/or pentyl groups yields the cations at m/z 234, 220, 214, 186 and 144. While the relative abundance of the ions varies among the six regioisomeric substances the 1-n-pentyl-3-benzoylindole and 1-n-pentyl-5-benzoylindole share very similar relative abundances for the major fragment ions. Chromatographic separations on a capillary column containing a 0.5μm film of 100% trifluoropropyl methyl polysiloxane (Rtx-200) provided excellent resolution of these six compounds. The elution order appears related to the relative distance between the two indole substituted groups. The latest eluting compounds (highest retention time) have the two substituents on opposite sides of the indole nucleus. Infrared absorption spectral data show the carbonyl absorption band for each of the benzoylindoles and provide distinguishing and characteristic information to individualize each of the regioisomers in this set of compounds.
Product Description
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard