Merck
  • An engineered old yellow enzyme that enables efficient synthesis of (4R,6R)-Actinol in a one-pot reduction system.
An engineered old yellow enzyme that enables efficient synthesis of (4R,6R)-Actinol in a one-pot reduction system.
Chembiochem : a European journal of chemical biology (2015-02-03)
Shoichiro Horita, Michihiko Kataoka, Nahoko Kitamura, Takuya Nakagawa, Takuya Miyakawa, Jun Ohtsuka, Koji Nagata, Sakayu Shimizu, Masaru Tanokura
ABSTRACT
(4R,6R)-Actinol can be stereo-selectively synthesized from ketoisophorone by a two-step conversion using a mixture of two enzymes: Candida macedoniensis old yellow enzyme (CmOYE) and Corynebacterium aquaticum (6R)-levodione reductase. However, (4S)-phorenol, an intermediate, accumulates because of the limited substrate range of CmOYE. To address this issue, we solved crystal structures of CmOYE in the presence and absence of a substrate analogue p-HBA, and introduced point mutations into the substrate-recognition loop. The most effective mutant (P295G) showed two- and 12-fold higher catalytic activities toward ketoisophorone and (4S)-phorenol, respectively, than the wild-type, and improved the yield of the two-step conversion from 67.2 to 90.1%. Our results demonstrate that the substrate range of an enzyme can be changed by introducing mutation(s) into a substrate-recognition loop. This method can be applied to the development of other favorable OYEs with different substrate preferences.
Product Description
USP
Menadione, United States Pharmacopeia (USP) Reference Standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
4-Oxoisophorone, ≥98%, FG
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%
Sigma-Aldrich
2,6,6-Trimethyl-2-cyclohexene-1,4-dione, 98%