Merck
  • Polypyrrole-montmorillonite nanocomposite as sorbent for solid-phase microextraction of phenolic compounds in water.
Polypyrrole-montmorillonite nanocomposite as sorbent for solid-phase microextraction of phenolic compounds in water.
Journal of separation science (2014-09-13)
Mir Mahdi Abolghasemi, Sheyda Parastari, Vahid Yousefi
ABSTRACT
A fiber-coated polypyrrole-montmorillonite nanocomposite was prepared for solid-phase microextraction. The fiber coating can be prepared easily; it is mechanically stable and exhibits relatively high thermal stability. The prepared fiber was evaluated for the extraction of some phenolic compounds from aqueous sample solutions by gas chromatography-mass spectrometry. The effects of the extraction and desorption parameters including extraction time, extraction temperature, stirring rate, ionic strength, pH and desorption temperature and time have been studied. At optimum conditions, the repeatability for one fiber (n = 5), expressed as % relative standard deviation was between 6.5 and 7.8% for the phenolic compounds. The detection limits for the studied phenolic compounds were between 0.05-1.3 ng/mL. The developed method offers the advantage of being simple to use, with shorter analysis time, lower cost, thermal stability of the fibers, and high relative recovery in comparison to conventional methods of analysis.
Sigma-Aldrich
2,6-Dichlorophenol, 99%
Sigma-Aldrich
4-Chlorophenol, ≥99%
Sigma-Aldrich
2,4-Dichlorophenol, 99%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%
Sigma-Aldrich
Phenol, natural, 97%, FG
USP
Phenol, United States Pharmacopeia (USP) Reference Standard