Merck
  • A robust smart window: reversibly switching from high transparency to angle-independent structural color display.
A robust smart window: reversibly switching from high transparency to angle-independent structural color display.
Advanced materials (Deerfield Beach, Fla.) (2015-03-04)
Dengteng Ge, Elaine Lee, Lili Yang, Yigil Cho, Min Li, Daniel S Gianola, Shu Yang
ABSTRACT
A smart window is fabricated from a composite consisting of elastomeric poly(dimethylsiloxane) embedded with a thin layer of quasi-amorphous silica nanoparticles. The smart window can be switched from the initial highly transparent state to opaqueness and displays angle-independent structural color via mechanical stretching. The switchable optical property can be fully recovered after 1000 stretching/releasing cycles.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%