Merck
  • Genome sequence and genome mining of a marine-derived antifungal bacterium Streptomyces sp. M10.
Genome sequence and genome mining of a marine-derived antifungal bacterium Streptomyces sp. M10.
Applied microbiology and biotechnology (2015-02-18)
Jingbo Tang, Xingyan Liu, Jian Peng, Yong Tang, Yang Zhang
ABSTRACT
A marine-derived actinobacteria Streptomyces sp. M10 was identified as a prolific antifungal compounds producer and shared a 99.02 % 16S ribosomal RNA (rRNA) sequence similarity with that of Streptomyces marokkonensis Ap1(T), which can produce polyene macrolides. To further evaluate its biosynthetic potential, the 7,207,169 bp Streptomyces sp. M10 linear chromosome was sequenced and mined for identifiable secondary metabolite-associated gene clusters. A total of 20 secondary metabolite-associated gene clusters were deduced, including three polyketide synthases (PKSs), four non-ribosomal peptide synthetases (NRPSs), four hybrid NRPS-PKSs, three NRPS-independent siderophores, and two lantibiotic and four terpene biosynthetic gene clusters. One of the type I PKS gene cluster, pks1, shared a 85 % nucleotide similarity with candicidin/FR008 gene cluster, indicating the capacity of this organism to produce polyene macrolides. This assumption was verified by a scale-up culturing of Streptomyces sp. M10 on A1 agar plates, which lead to the isolation of two polyene families PF1 and PF2, with characteristic UV adsorption at 269, 278, and 290 nm (PF1) and 363, 386, and 408 nm (PF2), respectively. Compound 9-04 was further purified from PF1, and its chemical structure was partially elucidated to be a typical polyene macrolide by NMR and UV spectrum. This study affirmatively identified Streptomyces sp. M10 as a source of polyene metabolites and highlighted genome mining of interested organism as a powerful tool for natural product discovery.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%