Merck
  • Ampelopsin suppresses breast carcinogenesis by inhibiting the mTOR signalling pathway.
Ampelopsin suppresses breast carcinogenesis by inhibiting the mTOR signalling pathway.
Carcinogenesis (2014-05-28)
Hui Chang, Xiaoli Peng, Qian Bai, Yong Zhou, Xiaoping Yu, Qianyong Zhang, Jundong Zhu, Mantian Mi
ABSTRACT
The mammalian target of rapamycin (mTOR), which is a master regulator of cellular catabolism and anabolism, plays an important role in tumourigenesis and progression. In this study, we report the chemopreventive effect of the dietary compound ampelopsin (AMP) on breast carcinogenesis in vivo and in vitro, which acts by inhibiting the mTOR signalling pathway. Our study indicates that AMP treatment effectively suppresses 1-methyl-1-nitrosourea (MNU)-induced breast carcinogenesis in rats and inhibits 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) and benzo[a]pyrene (B[a]P)-induced cellular carcinogenesis. Additionally, AMP inhibits the growth of breast cancer cells in vitro and in vivo. The activity of mTOR kinase was found to be significantly increased in a time-dependent manner during chronic breast carcinogenesis, and this increase can be suppressed by AMP co-treatment. AMP also effectively suppresses mTOR activity in breast cancer MDA-MB-231 cells. We also demonstrated that AMP is an effective mTOR inhibitor that binds to one site on the mTOR target in two ways. Further studies confirmed that AMP inhibits the activation of Akt, suppresses the formation of mTOR complexes (mTORC)1/2 by dissociating regulatory-associated protein of mTOR and rapamycin-insensitive companion of mTOR and, consequently, decreases the activation of the downstream targets of mTOR, including ribosomal p70-S6 kinase, ribosomal protein S6, eukaryotic translation initiation factor 4B and eukaryotic translation initiation factor 4E-binding protein 1. These finding suggest that AMP is a bioactive natural chemopreventive agent against breast carcinogenesis and is an effective mTOR inhibitor that may be developed as a useful chemotherapeutic agent in the treatment of breast cancer.
Product Description
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%