Merck
  • Simultaneous gas-chromatographic urinary measurement of sugar probes to assess intestinal permeability: use of time course analysis to optimize its use to assess regional gut permeability.
Simultaneous gas-chromatographic urinary measurement of sugar probes to assess intestinal permeability: use of time course analysis to optimize its use to assess regional gut permeability.
Clinica chimica acta; international journal of clinical chemistry (2015-01-17)
Maliha Shaikh, Kumar Rajan, Christopher B Forsyth, Robin M Voigt, Ali Keshavarzian
ABSTRACT
Measurement of intestinal permeability is important in several diseases but currently several methods are employed. We sought to: (1) develop a new GC based method to measure urinary mannitol, lactulose and sucralose to assess regional and total gut permeability; (2) analyze the kinetics of these sugars in the urine to determine which ratio is useful to represent intestinal permeability; and (3) determine whether age, gender, race and BMI impact these values. Subjects drank a cocktail of sucrose, lactulose, mannitol and sucralose and these sugars were measured in the urine at 5, 12 and 24h with gas chromatography. Urinary mannitol exhibited significantly different kinetics than lactulose and sucralose which were similar to each other and varied little over the 24h. No permeability differences were observed for renal function, age, race, sex, or BMI. Our data do not support the use of the widely used L/M ratio as an accurate estimate of intestinal permeability. Our data support the use of: the sucralose/lactulose (S/M) ratio to measure: small intestine permeability (first 5h); small and large intestine (first 12h), and total gut permeability (24h). This was also found to be true in a Parkinson's disease model.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Ammonium trifluoroacetate, 98%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone, analytical standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)