Merck
  • A widely used in vitro biofilm assay has questionable clinical significance for enterococcal endocarditis.
A widely used in vitro biofilm assay has questionable clinical significance for enterococcal endocarditis.
PloS one (2014-09-26)
Anne-Marie Leuck, James R Johnson, Gary M Dunny
ABSTRACT
Biofilm formation may play an important role in the pathogenesis of infections caused by Enterococcus faecalis, including endocarditis. Most biofilm studies use a polystyrene dish assay to quantify biofilm biomass. However, recent studies of E. faecalis strains in tissue and animal models suggest that polystyrene dish results need to be interpreted with caution. We evaluated 158 clinical E. faecalis isolates using a polystyrene dish assay and found variation in biofilm formation, with many isolates forming little biofilm even when different types of media were used. However, all tested clinical isolates were able to form biofilms on porcine heart valve explants. Dextrose-enhanced biofilm formation in the polystyrene dish assay was found in 6/12 (50%) of clinical isolates tested and may explain some, but not all of the differences between the polystyrene dish assay and the heart valve assay. These findings suggest that in studies assessing the clinical relevance of enterococcal biofilm-forming ability, ex vivo biofilm formation on a relevant tissue surface may be warranted to validate results of in vitro assays.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 60 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 3 mg substrate per tablet
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications