Merck
  • Quantitative assessment of intestinal first-pass metabolism of oral drugs using portal-vein cannulated rats.
Quantitative assessment of intestinal first-pass metabolism of oral drugs using portal-vein cannulated rats.
Pharmaceutical research (2014-08-29)
Yoshiki Matsuda, Yoshihiro Konno, Takashi Hashimoto, Mika Nagai, Takayuki Taguchi, Masahiro Satsukawa, Shinji Yamashita
ABSTRACT
To evaluate the impact of intestinal first-pass metabolism (Fg) by cytochrome P4503A (CYP3A) and uridine 5'-diphosphate-glucuronosyltransferases (UGT) on in vivo oral absorption of their substrate drugs. CYP3A and UGT substrates were orally administered to portal-vein cannulated (PV) rats to evaluate their intestinal availability (Fa · Fg). In the case of CYP3A substrates, vehicle or 1-aminobenzotriazole (ABT), a potent inhibitor of CYP enzymes, was pretreated to assess Fg separately from Fa (Enzyme-inhibition method). On the other hand, since potent inhibitors of UGT have not been identified, Fg of UGT substrate was calculated from total amount of metabolites generated in enterocytes (Metabolite-distribution method). After oral administration of CYP3A substrates in ABT-pretreated rats, the portal and systemic plasma concentrations of the metabolite were nearly the same, indicating almost complete inhibition of intestinal CYP3A-mediated metabolism. Using Enzyme-inhibition method, Fg of midazolam (1 mg/kg) was calculated as 0.71. Additionally, total amount of raloxifene-6-glucuronide generated in enterocytes after oral administration of raloxifene was estimated using Metabolite-distribution method and Fg of raloxifene (0.98 μmol/kg) was calculated as 0.21. PV rats enabled in vivo quantitative assessment of intestinal first-pass metabolism by CYP3A and UGT. This method is useful for clarifying the cause of low bioavailability.
Sigma-Aldrich
Felodipine, solid
Felodipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aminobenztropine, solid
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
1-Aminobenzotriazole
Supelco
Antipyrine, analytical standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material