Merck
  • Endometriosis also affects the decidua in contact with the fetal membranes during pregnancy.
Endometriosis also affects the decidua in contact with the fetal membranes during pregnancy.
Human reproduction (Oxford, England) (2014-11-30)
L Marcellin, P Santulli, J Gogusev, C Lesaffre, S Jacques, C Chapron, F Goffinet, D Vaiman, C Méhats
ABSTRACT
Are the fetal membranes of women affected with endometriosis similar to those from disease-free women? Decidua of women with endometriosis is able to generate endometriotic-like lesions in contact with the fetal membranes. Eutopic endometrium of women affected with endometriosis presents compromised properties. Endometrium undergoes decidualisation to accept and to further control the conceptus development during pregnancy. Decidualized endometrium is in close contact with the chorionic membrane and forms the choriodecidual layer, a major maternal-fetal interface. This is a laboratory case-control study involving diseased versus control samples. Eleven case samples and 11 control samples were collected from women in a tertiary care/research center between November 2011 and December 2013. Participants were consecutive pregnant women affected with confirmed endometriosis and disease free women, who underwent Cesarean section before labor for obstetrical indication. The choriodecidual tissues were characterized using histology, immunohistochemistry, transcriptomic and whole genome CpG methylation analyses. We demonstrate for the first time the presence of endometriotic-like lesions within the decidual side of the choriodecidua of the fetal membranes from women affected with severe endometriosis. Fetal membranes from women affected with endometriosis exhibited glandular components in the choriodecidual layer surrounded by enlarged decidualized cells disseminated along the entire membrane surface. Significant deregulation (variation of expression ≥2, P-value ≤0.05) was observed for 2773 genes known to be enriched in processes involved in glandular function, endocrine and nervous system, neoangiogenesis, and autoimmune disease. CpG methylation analysis revealed 5999 differentially methylated regions with a P-value ≤0.05. We studied women who delivered at term by Cesarean section before labor, following an uneventful pregnancy. Notwithstanding this, one cannot exclude that the presence of disseminated endometriotic lesions within the choriodecidual layer of the fetal membranes may disturb the anatomical integrity and/or the function of the membranes in some women with endometriosis. Our results shed new light on the capability of the diseased decidua to develop lesions not only at ectopic autologous locations, but also on the semi-allogenous fetal membranes, a particularly immunotolerant environment.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
o-Xylene, analytical standard
Supelco
o-Xylene, suitable for HPLC, 98%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications