Merck
  • Medroxyprogesterone acetate enhances monocyte-endothelial interaction under flow conditions by stimulating the expression of cell adhesion molecules.
Medroxyprogesterone acetate enhances monocyte-endothelial interaction under flow conditions by stimulating the expression of cell adhesion molecules.
The Journal of clinical endocrinology and metabolism (2014-03-13)
Fumitake Ito, Hiroshi Tatsumi, Taisuke Mori, Izumi Suganuma, Yukiko Tanaka, Aya Sasaki, Seiki Matsuo, Koichi Iwasa, Jo Kitawaki
ABSTRACT
Monocyte adhesion to endothelial cells is an important initial event in atherosclerosis and is partially mediated by adhesion molecule expression on the cell surface. Although estrogens inhibit atherosclerosis development, effects of coadministered progestogen remain controversial. We examined the effects of progestogen on cytokine-stimulated human umbilical venous endothelial cell (HUVEC) expression of adhesion molecules. In HUVECs, adhesion molecule mRNA levels were measured by real-time PCR. Protein expression was quantified by immunocytochemistry and ELISAs. To mimic the monocyte adherence to endothelial cells, we used a flow chamber system to assess progestogen effects on U937 monocytoid cell adherence to HUVEC monolayers. We also examined the suppression effects of adhesion molecules with small interference RNAs. mRNA levels of adhesion molecules in HUVECs treated with medroxyprogesterone acetate (MPA) or 17β-estradiol + MPA were 1.7- to 2.5-fold higher than those in the control. MPA increased the protein expression of E-selectin, P-selectin, and intercellular adhesion molecule-1 compared with that for the control (83.0 ± 0.7, 34.8 ± 1.2, and 5.4 ± 0.0 ng/mL, respectively), whereas other progestogens or 17β-estradiol additive to progestogens did not significantly change expression. MPA significantly increased U937 monocytoid cell adherence compared with the control (56.0 ± 1.5 vs 46.5 ± 3.5 adherent cells per 10 fields) but did not increase adherence to HUVECs with knocked down intercellular adhesion molecule-1. MPA increases cell adhesion molecule expression on HUVECs, causing increased numbers of monocytoid cells to adhere to HUVECs. These MPA effects may be a risk factor for atherogenesis on endothelial cells in postmenopausal women receiving hormone replacement therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Icam1
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human ICAM1
Sigma-Aldrich
Hydroxyflutamide, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O