Merck
  • Orexin type 1 receptor antagonism in Lateral Paragigantocellularis nucleus attenuates naloxone precipitated morphine withdrawal symptoms in rats.
Orexin type 1 receptor antagonism in Lateral Paragigantocellularis nucleus attenuates naloxone precipitated morphine withdrawal symptoms in rats.
Neuroscience letters (2013-11-12)
S Mohammad Ahmadi-Soleimani, Masoumeh Ghaemi-Jandabi, Hossein Azizi, Saeed Semnanian
ABSTRACT
Orexin neuropeptides have been reported to be involved in morphine induced physical dependence and withdrawal. The Lateral Paragigantocellularis (LPGi) is a key brain region implicated in the expression of somatic signs of morphine withdrawal syndrome. Orexin A and orexin type 1 receptor have been found in LPGi neurons but the effect of orexin on the expression of opiate dependence and withdrawal phenomena in this brain structure has not been studied yet. In this study, the effect of intra-LPGi administration of SB 334867 (selective orexin type 1 receptor antagonist) on the behavioral signs of morphine withdrawal syndrome was investigated. Male Wistar rats weighing 250-300 g were rendered dependent by adding morphine sulfate (Temad, Tehran, Iran) to their drinking water in increasing concentrations of 0.1, 0.2, 0.3mg/ml for every 48 h and 0.4 mg/ml during the next 15 days. Behavioral signs of morphine withdrawal were assessed in a transparent cylindrical Plexiglas test chamber (30 cm diameter, 50 cm height) for 25 min. One group of animals received intra-LPGi injection of SB 334867 (0.2 μl, 100 μM) immediately before naloxone. In the control group, SB-334867 vehicle (DMSO 1%, 0.2 μl) was microinjected into LPGi. Our results indicate that intra-LPGi administration of SB 334867 significantly decreases naloxone precipitated morphine withdrawal signs. Thus, it seems that orexin might have a pivotal role in the expression of morphine withdrawal signs through affecting orexin type 1 receptor in LPGi nucleus.
Product Description
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%