Merck
  • Surface-enhanced fluorescence immunosensor using Au nano-crosses for the detection of microcystin-LR.
Surface-enhanced fluorescence immunosensor using Au nano-crosses for the detection of microcystin-LR.
Biosensors & bioelectronics (2014-07-14)
Yun Li, Jiadi Sun, Longyun Wu, Jian Ji, Xiulan Sun, Yongzhong Qian
ABSTRACT
A surface-enhanced fluorescence (SEF) immunosensor for the detection of microcystin-LR was developed using Au nano-crosses as fluorescence enhancement nanoparticles and cy5 as a fluorescence label molecule. The SEF effects of cy5 in the proximity of Au nanorods and gold nano-crosses was investigated by using Au nanorods or nano-crosses coated negative-charged glass surfaces. Fluorescence measurements indicated that SEF was influenced by the size, shape and distribution of the Au nanoparticles, with an appropriate spacer layer between the Au nanoparticles and the cy5. The enhancement factor was from 2.3- to 35-fold. Under optimal conditions, the SEF immunosensor exhibited a good linear response at microcystin-LR concentrations of 0.02-16 ng mL(-1) (R(2)=0.9981). The limit of detection was 0.007 ng mL(-1) with little adsorption of microcystin-RR, microcystin-LW, and microcystin-LF. High microcystin-LR recoveries were obtained from naturally contaminated fish samples. The SEF immunosensor allows the reliable detection of microcystin-LR in seafood, and has potential in simple, sensitive detection applications.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
1-Dodecanethiol, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
5-Bromosalicylic acid, technical grade, 90%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications