Merck
  • A HPLC-SRM-MS based method for the detection and quantification of methotrexate in urine at doses used in clinical practice for patients with rheumatological disease: a potential measure of adherence.
A HPLC-SRM-MS based method for the detection and quantification of methotrexate in urine at doses used in clinical practice for patients with rheumatological disease: a potential measure of adherence.
The Analyst (2015-02-12)
J Bluett, I Riba-Garcia, K Hollywood, S M M Verstappen, A Barton, R D Unwin
ABSTRACT
Rheumatoid arthritis (RA) is a common autoimmune disease that causes significant disability and reduced life expectancy. The folate antagonist methotrexate (MTX) is first-line therapy for RA when used weekly at low doses (5-25 mg). However, the true rate of adherence to MTX is uncertain. This is in part due to the different methods of measurement of adherence employed with no biochemical test currently available to determine adherence to low dose MTX. Common methods of MTX measurement include immunoassays in patients with high dose therapy, but these assays cross-react with MTX metabolites and lack the sensitivity required to measure adherence to low dose MTX. HPLC-SRM-MS (selected reaction monitoring-mass spectrometry) has several theoretical advantages over immunoassays with improved specificity, minimal cross-reaction and higher sensitivity. The aim of this study was to develop an assay to measure MTX and its major metabolite 7-OH-MTX in urine as a tool to monitor adherence to low dose MTX in clinic. As a proof of concept, urine samples from 4 participants with RA were measured after directly observed therapy. The assay showed improved sensitivity compared to that reported by immunoassays, with low carryover and high within-run precision. In participant samples, MTX was measurable in the urine for up to 105 hours after administration and 7-OH-MTX was detectable up to 98 hours after administration, suggesting that this assay is suitable for the measurement of adherence to therapy. The assay requires minimal sample preparation and can be adopted by other laboratories with minimal study set up.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)