Merck
  • In vitro antibacterial phenolic extracts from "sugarbag" pot-honeys of Australian stingless bees (Tetragonula carbonaria).
In vitro antibacterial phenolic extracts from "sugarbag" pot-honeys of Australian stingless bees (Tetragonula carbonaria).
Journal of agricultural and food chemistry (2014-11-26)
C Flavia Massaro, Daniel Shelley, Tim A Heard, Peter Brooks
ABSTRACT
Australian stingless bee honeys have been shown to exert antioxidant and in vitro antimicrobial properties; however their bioactive factors remained unidentified. This study investigated the antibacterial properties of phenolic extracts from Tetragonula carbonaria honeys. Honeys were harvested from beehives in three sites of South East Australia. Liquid-liquid extractions yielded the phenolic concentrates, for analyses by liquid and gas chromatography mass spectrometry. Antibacterial assays were conducted against Staphylococcus aureus and Klebsiella pneumoniae by in vitro agar diffusion and broth dilution assays. The phenolic extracts averaged to 5.87 mg/100 g of raw honeys, and constituents were 3-phenyllactic acid, lumichrome, diglycosylflavonoids, norisoprenoids. The honeys did not contain methylglyoxal, dihydroxyacetone or phenolics characteristic of Leptospermum nectars. Hydrogen peroxide content amounted up to 155.8 μM in honeys. Beside the bactericidal effects of hydrogen peroxide at 760 μM, other antibacterial factors were the phenolic extracts of "sugarbag" honeys that were active at minimum bactericidal concentrations of 1.2-1.8 mg/mL.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
5-(Hydroxymethyl)furfural, analytical standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Methanol, analytical standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Supelco
Hexadecane, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hydroxyacetone, contains ≤500 ppm sodium carbonate as stabilizer, technical grade, 90%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
3-Phenyllactic acid, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%