Merck
  • Synthesis and biological evaluation of carbon-11 and fluorine-18 labeled tracers for in vivo visualization of PDE10A.
Synthesis and biological evaluation of carbon-11 and fluorine-18 labeled tracers for in vivo visualization of PDE10A.
Nuclear medicine and biology (2014-07-09)
Maarten Ooms, Sofie Celen, Michel Koole, Xavier Langlois, Mark Schmidt, Meri De Angelis, José Ignacio Andrés, Alfons Verbruggen, Koen Van Laere, Guy Bormans
ABSTRACT
In vivo visualization of PDE10A using PET provides a tool to evaluate the role of PDE10A in various neuropsychiatric diseases and can also be useful in the clinical evaluation of PDE10A inhibitor drug candidates. We evaluated several carbon-11 and fluorine-18 labeled PDE10A inhibitors as potential PDE10A PET radioligands. [(11)C]MP10, [(11)C]JNJ42071965 and four other tracers were developed. Their biodistribution was evaluated in rats. Rat plasma and brain radiometabolites were quantified. Baseline microPET imaging was performed in normal rats and PDE10A knockout (KO) and wild-type (WT) mice. Blocking and displacement studies were conducted. The selectivity of the tracer binding was further studied in an ex vivo autoradiography experiment in PDE10A KO and WT mice. Biodistribution showed brain uptake for all tracers in the striatum and wash-out from the cerebellum. [(11)C]1 ((11)C-MP10) had the highest specific uptake index (striatum (S) vs. cerebellum (C) ratios (S/C)-1) at 60 min (7.4). [(11)C]5 ([(11)C]JNJ42071965) had a high index at the early time points (1.0 and 3.7 at 2 and 30 min p.i., respectively). The affinity of [(11)C]4, [(18)F]3 and [(18)F]6 was too low to visualize PDE10A using microPET. [(11)C] 2 showed a specific binding, while kinetics of [(11)C]1 were too slow. [(11)C]5 reached equilibrium after 10 min (uptake index=1.2). Blocking and displacement experiments in rats and baseline imaging in PDE10A KO mice showed specific and reversible binding of [(11)C]5 to PDE10A. We successfully radiolabeled and evaluated six radiotracers for their potential to visualize PDE10A in vivo. While [(11)C]1 had the highest striatal specific uptake index, its slow kinetics likely compromise clinical use of this tracer. [(11)C]5 has a relatively high striatum-to-background ratio and fast kinetic profile, which makes it a valuable carbon-11 alternative.
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%