Merck
  • Assessing non-digestible compounds in apple cultivars and their potential as modulators of obese faecal microbiota in vitro.
Assessing non-digestible compounds in apple cultivars and their potential as modulators of obese faecal microbiota in vitro.
Food chemistry (2014-05-20)
Luis Condezo-Hoyos, Indira P Mohanty, Giuliana D Noratto
ABSTRACT
The health benefits of apple bioactive compounds have been extensively reported. However, only few studies have focused on bioactive compounds that are not absorbed and metabolised during gastrointestinal digestion and can induce changes in microbial populations of faeces. We have characterised Braeburn, Fuji, Gala, Golden Delicious, Granny Smith, McIntosh and Red Delicious cultivars and found significant differences for extractable phenolics (1.08-9.2mg/g) non-extractable proanthocyanidins (3.28-5.7mg/g), and dietary fibre (20.6-32.2%) among cultivars with Granny Smith having the highest contents. Granny Smith was used after in vitro digestion for fermentation of faeces from diet-induced obese mice. Results showed that relative abundances of Firmicutes, Bacteroidetes, Enterococcus, Enterobacteriaceae, Escherichia coli, and Bifidobacterium in apple cultured faeces tended to resemble the abundance in faeces from lean mice with increased trend in the production of butyric acid. These results suggest that apple non-digestible compounds might help to re-establish a disturbed microbiota balance in obesity.
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Supelco
Glycerol, analytical standard
SAFC
Tromethamine
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, analytical standard
Supelco
Methanol, analytical standard
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications