Merck
  • Seed oil of Joannesia princeps improves cutaneous wound closure in experimental mice.
Seed oil of Joannesia princeps improves cutaneous wound closure in experimental mice.
Acta histochemica (2014-07-24)
Aline Donato-Trancoso, Lenicio Gonçalves, Andréa Monte-Alto-Costa, Francisco de Assis da Silva, Bruna Romana-Souza
ABSTRACT
Joannesia princeps (Cotieira) is a well known medicinal plant in Brazil, however, the therapeutic effects of oil obtained from its seeds have still not been demonstrated. The beneficial effects of J. princeps seed oil on cutaneous wound healing on the back of experimental mice were investigated. An excisional lesion in male Swiss mice (n=20 per group) was topically treated with mineral oil or J. princeps seed oil once a day beginning on the day of lesion until the third day after wounding. Animals were killed and lesions collected after 14 days. Murine skin fibroblast cultures were treated with J. princeps seed oil and fibroblast activity was evaluated. In the in vivo assay, J. princeps seed oil increased wound contraction and migratory tongue length, but reduced neutrophil and macrophage number when compared with the control group. Blood vessel number, collagen deposition, and VEGF levels were increased in treated lesions when compared with control lesions. However, J. princeps seed oil reduced myofibroblast density and carbonyl protein levels when compared with the control group. In the in vitro assay, treatment with J. princeps seed oil increased fibroblast migration and proliferation, but reduced myofibroblastic differentiation in vitro. In conclusion, J. princeps seed oil accelerates wound closure increasing angiogenesis, keratinocyte migration, and fibroblast activity while reducing inflammatory response and oxidative damage.
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Supelco
4-(Dimethylamino)benzaldehyde solution, 10 g/L in isopropanol, for TLC derivatization
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N