Merck
  • Nitric oxide donors reduce the invasion ability of ovarian cancer cells in vitro.
Nitric oxide donors reduce the invasion ability of ovarian cancer cells in vitro.
Anti-cancer drugs (2014-07-19)
Michal Kielbik, Izabela Szulc, Marta Brzezinska, Katarzyna Bednarska, Patrycja Przygodzka, Zofia Sulowska, Marek Nowak, Magdalena Klink
ABSTRACT
The most important factors involved in tumor metastasis and angiogenesis are metalloproteinases (MMPs), vascular endothelial growth factor, and multifunctional transforming growth factor β1. These factors are responsible for extracellular matrix degradation, induction of vascular permeability, and enhancement of tumor cells' invasion and metastasis. Elevated expression and secretion of the above-mentioned factors are correlated with the higher aggressiveness of tumors and low patient survival for example, patients with ovarian cancer. Therefore, regulation of the expression, secretion, and activity of these factors is still considered a potent target for therapeutic intervention in cancer patients. Nitric oxide (NO) donors belong to the class of agents with multivalent targeted activities in cancer cells and are considered potential anticancer therapeutics. Our studies have shown that NO donors such as spermine/NO and diethylenetriamine/NO decrease the secretion of vascular endothelial growth factor-A from the OVCAR-3 ovarian cancer cell line, but not from the SK-OV-3 ovarian cancer cell line. The release of MMP-2 from both cell lines was reduced in a soluble guanylate cyclase-dependent manner by spermine/NO and diethylenetriamine/NO. Nevertheless, MMP-2 activity was only affected in SK-OV-3 cells. Both NO donors reduced the transmigration of the ovarian cancer cell lines. We did not observe any significant effect of spermine/NO and diethylenetriamine/NO on mRNA expression of the tested aggressiveness factors. In conclusion, our data indicated that NO donors reduced the metastatic potential of ovarian cancer cells, but its impact is rather low and requires high concentrations of donors. Moreover, both the tested cell lines differed in the susceptibility to NO donors.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder