Merck
  • Evaluation of a low-density hydrogel microarray technique for mycobacterial species identification.
Evaluation of a low-density hydrogel microarray technique for mycobacterial species identification.
Journal of clinical microbiology (2015-01-23)
Danila V Zimenkov, Elena V Kulagina, Olga V Antonova, Maria A Krasnova, Ekaterina N Chernyaeva, Vyacheslav Y Zhuravlev, Alexey V Kuz'min, Sergey A Popov, Alexander S Zasedatelev, Dmitry A Gryadunov
ABSTRACT
In addition to the obligatory pathogenic species of the Mycobacterium tuberculosis complex and Mycobacterium leprae, the genus Mycobacterium also includes conditionally pathogenic species that in rare cases can lead to the development of nontuberculous mycobacterial diseases. Because tuberculosis and mycobacteriosis have similar clinical signs, the accurate identification of the causative agent in a clinical microbiology laboratory is important for diagnostic verification and appropriate treatment. This report describes a low-density hydrogel-based microarray containing oligonucleotide probes based on the species-specific sequences of the gyrB gene fragment for mycobacterial species identification. The procedure included the amplification of a 352-nucleotide fragment of the gene and its hybridization on a microarray. The triple-species-specific probe design and the algorithm for hybridization profile recognition based on the calculation of Pearson correlation coefficients, followed by the construction of a profile database, allowed for the reliable and accurate identification of mycobacterial species, including mixed-DNA samples. The assay was used to evaluate 543 clinical isolates from two regions of Russia, demonstrating its ability to detect 35 mycobacterial species, with 99.8% sensitivity and 100% specificity when using gyrB, 16S, and internal transcribed spacer (ITS) fragment sequencing as the standard. The testing of clinical samples showed that the sensitivity of the assay was 89% to 95% for smear-positive samples and 36% for smear-negative samples. The large number of identified species, the high level of sensitivity, the ability to detect mycobacteria in clinical samples, and the up-to-date profile database make the assay suitable for use in routine laboratory practice.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
trans-Sinapic acid, analytical standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O