Merck
  • Galectin-3 knockdown increases gefitinib sensitivity to the inhibition of EGFR endocytosis in gefitinib-insensitive esophageal squamous cancer cells.
Galectin-3 knockdown increases gefitinib sensitivity to the inhibition of EGFR endocytosis in gefitinib-insensitive esophageal squamous cancer cells.
Medical oncology (Northwood, London, England) (2015-03-20)
Guanghui Cui, Mingwei Cui, Yuhang Li, Youguang Liang, Weihao Li, Haizhou Guo, Song Zhao
ABSTRACT
Esophageal cancer (EC) is one of the most aggressive malignancies with a distinctly high incidence and mortality rate. Esophageal squamous cell carcinoma (ESCC) is the major histologic subtype of EC, with 40-70 % of tumors overexpressing the epidermal growth factor receptor (EGFR). Blockade of EGFR signal transduction may be a promising and effective strategy for EC therapy. However, the therapeutic efficacy of EGFR-tyrosine kinase inhibitors is clinically limited because of drug resistance. Galectin-3, a member of the animal lectin family, has been associated with a variety of biological functions and the progression of multiple tumors, including ESCC. In this study, we investigated the role of galectin-3 involved in potential gefitinib-resistance mechanisms in EGFR-positive ESCC cell lines. The results revealed that gefitinib treatment induced different inhibitory effects on cell viability, cell cycle progression and cell invasion in gefitinib-sensitive KYSE-450 and gefitinib-insensitive TE-8 cells with different levels of galectin-3 expression. Interestingly, we further found that EGF-induced EGFR endocytosis and EGFR signaling were different between gefitinib-sensitive and gefitinib-insensitive ESCC cell lines. Galectin-3 inhibition in combination with gefitinib treatment induced greater inhibitory effects on cell viability, cell cycle progression and cell invasion in gefitinib-insensitive TE-8 cells. Moreover, galectin-3 inhibition increased the gefitinib sensitivity of TE-8 cells in terms of EGFR endocytosis in vitro and anti-tumor effects in vivo. Taken together, galectin-3 knockdown increased gefitinib sensitivity through the inhibition of EGFR endocytosis in gefitinib-insensitive ESCC cells and galectin-3 may be a rational therapeutic target in ESCC with gefitinib resistance.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%