Merck
  • Regulation of later neurogenic stages of adult-derived neural stem/progenitor cells by L-type Ca2+ channels.
Regulation of later neurogenic stages of adult-derived neural stem/progenitor cells by L-type Ca2+ channels.
Development, growth & differentiation (2014-10-07)
Daniel B L Teh, Toru Ishizuka, Hiromu Yawo
ABSTRACT
In the adult hippocampus, new neurons are continuously generated and incorporated into the local circuitry in a manner dependent on the network activity. Depolarization evoked by neurotransmitters has been assumed to activate L-type Ca2+ channels (LTCC) which regulate the intracellular Ca2+ -dependent signaling cascades. The process of neurogenesis contains several stages such as proliferation, fate determination, selective death/survival and maturation. Here, we investigated which stage of neurogenesis is under the regulation of LTCC using a clonal line of neural stem/progenitor cells, PZ5, which was derived from adult rat hippocampus. Although undifferentiated PZ5 cells were type 1-like cells expressing both nestin and glial fibrillary acidic protein, they generated neuronal, astrocytic and oligodendrocytic populations in differentiation medium containing retinoic acid. Proliferation of undifferentiated PZ5 cells was dependent on neither the LTCC antagonist, nimodipine (Nimo) nor the LTCC agonists, Bay K 8644 (BayK) or FPL 64176 (FPL), whereas the fraction of neuronal population that expressed both βIII-tubulin and MAP2 was reduced by Nimo but increased by BayK or FPL. At an earlier period of differentiation (e.g., day 4), the fraction of PZ5 cells expressing HuC/D, pan-neuronal marker, was not affected either by the LTCC activation or inhibition. At a later period of differentiation (e.g., day 9), the fraction of dying neurons was decreased by LTCC activation and increased by LTCC inhibition. It is suggested that the LTCC activation facilitates the survival and maturation of immature neurons, and that its inhibition facilitates the neuronal death.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Nimodipine
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Nimodipine, Pharmaceutical Secondary Standard; Certified Reference Material
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Nimodipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water