Merck
  • Epac is required for GLP-1R-mediated inhibition of oxidative stress and apoptosis in cardiomyocytes.
Epac is required for GLP-1R-mediated inhibition of oxidative stress and apoptosis in cardiomyocytes.
Molecular endocrinology (Baltimore, Md.) (2015-02-27)
Supachoke Mangmool, Piriya Hemplueksa, Warisara Parichatikanond, Nipon Chattipakorn
ABSTRACT
Although the cardioprotective effects of glucagon-like peptide-1 and its analogs have been reported, the exact mechanisms of the glucagon-like peptide-1 receptor (GLP-1R) signaling pathway in the heart are still unclear. Activation of the GLP-1R has been shown to increase cAMP levels, thus eliciting protein kinase A- and exchange protein activated by cAMP (Epac)-dependent signaling pathways in pancreatic β-cells. However, which pathway plays an important role in the antioxidant and antiapoptotic effects of GLP-1R activation in the heart is not known. In this study, we demonstrated that stimulation of GLP-1Rs with exendin-4 attenuated H2O2-induced reactive oxygen species production and increased the synthesis of antioxidant enzymes, catalase, glutathione peroxidase-1, and manganese superoxide dismutase that is dependent on Epac. Additionally, exendin-4 has an antiapoptotic effect by decreasing a number of apoptotic cells, inhibiting caspase-3 activity, and enhancing the expression of antiapoptotic protein B-cell lymphoma 2, which is mediated through both protein kinase A- and Epac-dependent pathways. These data indicate a critical role for Epac in GLP-1R-mediated cardioprotection.
Sigma-Aldrich
N-Benzoyladenosine, 96%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Selenium, pellets, < 5mm, ≥99.999%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Glutathione, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Forskolin, For use in molecular biology applications