Merck
  • Identification and characterization of membrane androgen receptors in the ZIP9 zinc transporter subfamily: I. Discovery in female atlantic croaker and evidence ZIP9 mediates testosterone-induced apoptosis of ovarian follicle cells.
Identification and characterization of membrane androgen receptors in the ZIP9 zinc transporter subfamily: I. Discovery in female atlantic croaker and evidence ZIP9 mediates testosterone-induced apoptosis of ovarian follicle cells.
Endocrinology (2014-07-12)
A Håkan Berg, Charles D Rice, Md Saydur Rahman, Jing Dong, Peter Thomas
ABSTRACT
Rapid, cell surface-initiated, pregenomic androgen actions have been described in various vertebrate cells, but the receptors mediating these actions remain unidentified. We report here the cloning and expression of a cDNA from Atlantic croaker (Micropogonias undulatus) ovaries encoding a 33-kDa, seven-transmembrane protein with binding and signaling characteristics of a membrane androgen receptor that is unrelated to any previously described steroid receptor. Instead, croaker membrane androgen receptor has 81-93% amino acid sequence identity with zinc transporter ZIP9 (SLC39A9) subfamily members, indicating it is a ZIP9 protein. Croaker ZIP9 is expressed in gonadal tissues and in brain and is up-regulated in the ovary by reproductive hormones. Croaker ZIP9 protein is localized to plasma membranes of croaker granulosa cells and human breast cancer (SKBR-3) cells stably transfected with ZIP9. Recombinant croaker ZIP9 has a high affinity (dissociation constant, Kd, 12.7 nM), limited capacity (maximal binding capacity 2.8 nM/mg protein), displaceable, single binding site-specific for androgens, characteristic of steroid receptors. Testosterone activates a stimulatory G protein coupled to ZIP9, resulting in increased cAMP production. Testosterone promotes serum starvation-induced cell death and apoptosis in transfected cells and in croaker ovarian follicle cells that is associated with rapid increases in intracellular free zinc concentrations, suggesting an involvement of zinc in this nonclassical androgen action to promote apoptosis. These responses to testosterone are abrogated by treatment with ZIP9 small interfering RNA. The results provide the first evidence that zinc transporter proteins can function as specific steroid membrane receptors and indicate a previously unrecognized signaling pathway mediated by steroid receptors involving alterations in intracellular zinc.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b8
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b3
Sigma-Aldrich
MISSION® esiRNA, targeting human AREG
Sigma-Aldrich
MISSION® esiRNA, targeting human AKR1B1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Areg
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fdxr
Sigma-Aldrich
MISSION® esiRNA, targeting human FDXR
Sigma-Aldrich
MISSION® esiRNA, targeting human AR