Merck
  • Self-aggregated nanoparticles of carboxylic curdlan-deoxycholic acid conjugates as a carrier of doxorubicin.
Self-aggregated nanoparticles of carboxylic curdlan-deoxycholic acid conjugates as a carrier of doxorubicin.
International journal of biological macromolecules (2014-09-07)
Jing-Kun Yan, Hai-Le Ma, Xia Chen, Juan-Juan Pei, Zhen-Bin Wang, Jian-Yong Wu
ABSTRACT
In this study, a new non-toxic, biodegradable, biocompatible and water-soluble carboxylic curdlan bearing the dissociable COOH group in 100% purity, which was prepared by 4-acetamido-TEMPO-mediated oxidation, was hydrophobically modified by deoxycholic acid (DOCA) to attain novel amphiphilic curdlan derivatives (CCDs) for the preparation of nano-carriers for antitumor drug doxorubicin (DOX). Under the effect of ultrasonication, the carboxylic curdlan derivatives in water were self-aggregated into spherical nanoparticles with diameters ranging from 214 nm to 380 nm. The critical aggregation concentrations decreased from 0.047 mg/mL to 0.016 mg/mL with increasing DS of DOCA. DOX-loaded CCD nanoparticles were prepared in an aqueous medium with dialysis method. The DOX-CCD nanoparticles exhibited pH- and dose-dependent drug release profiles during in vitro release experiments. Moreover, the drug transport mechanism was Fickian diffusion according to the Ritger-Peppas model. The CCD nanoparticles might be explored as potential carriers for hydrophobic drugs with controlled release and delivery functions.
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 10-15 %
Sigma-Aldrich
Sodium hypochlorite solution, purum, ~10% (RT)
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 4.00-4.99 %
Sigma-Aldrich
Sodium chlorite, puriss. p.a., 80% (RT)
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chlorite, technical grade, 80%
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%