Merck
  • Recognitive nano-thin-film composite beads for the enantiomeric resolution of the metastatic breast cancer drug aminoglutethimide.
Recognitive nano-thin-film composite beads for the enantiomeric resolution of the metastatic breast cancer drug aminoglutethimide.
Journal of chromatography. A (2014-07-22)
Porkodi Kadhirvel, Manuel Azenha, Eric Schillinger, Mahadeo R Halhalli, António F Silva, Börje Sellergren
ABSTRACT
Straightforward crushing and sieving bulk polymeric R-aminoglutethimide-imprinted materials were prepared by classical free radical polymerization, whereas nano thin walled grafted imprinted materials were prepared using RAFT mediated control polymerization technique. A stoichiometric non-covalent approach based on a triply hydrogen bonding functional monomer-template 1:1 complex (K=599mol(-1)L(-1)) led to chiral selectors far outperforming previously used selectors for resolving this racemate. The recognitive materials produced here (enantioselectivity factors, α∼10) also have no match within the previously reported enantioselective imprinted polymers (α 1.2-4.5). We here demonstrate a potentially generic solution to produce good quality grafted MIPs for templates interacting by hydrogen bonding alone, relying on solvent polarity tuning, significantly extending the range of templates compatible with this format.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
2,3-Diaminopyridine, 95%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Chloroform, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material