Merck
  • Design and development of novel mitochondrial targeted nanocarriers, DQAsomes for curcumin inhalation.
Design and development of novel mitochondrial targeted nanocarriers, DQAsomes for curcumin inhalation.
Molecular pharmaceutics (2014-05-24)
Špela Zupančič, Petra Kocbek, M Gulrez Zariwala, Derek Renshaw, Mine Orlu Gul, Zeeneh Elsaid, Kevin M G Taylor, Satyanarayana Somavarapu
ABSTRACT
Curcumin has potent antioxidant and anti-inflammatory properties but poor absorption following oral administration owing to its low aqueous solubility. Development of novel formulations to improve its in vivo efficacy is therefore challenging. In this study, formulation of curcumin-loaded DQAsomes (vesicles formed from the amphiphile, dequalinium) for pulmonary delivery is presented for the first time. The vesicles demonstrated mean hydrodynamic diameters between 170 and 200 nm, with a ζ potential of approximately +50 mV, high drug loading (up to 61%) and encapsulation efficiency (90%), resulting in enhanced curcumin aqueous solubility. Curcumin encapsulation in DQAsomes in the amorphous state was confirmed by X-ray diffraction and differential scanning calorimetry analysis. The existence of hydrogen bonds and cation-π interaction between curcumin and vesicle building blocks, namely dequalinium molecules, were shown in lyophilized DQAsomes using FT-IR analysis. Encapsulation of curcumin in DQAsomes enhanced the antioxidant activity of curcumin compared to free curcumin. DQAsome dispersion was successfully nebulized with the majority of the delivered dose deposited in the second stage of the twin-stage impinger. The vesicles showed potential for mitochondrial targeting. Curcumin-loaded DQAsomes thus represent a promising inhalation formulation with improved stability characteristics and mitochondrial targeting ability, indicating a novel approach for efficient curcumin delivery for effective treatment of acute lung injury and the rationale for future in vivo studies.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
MES monohydrate, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Methanol, analytical standard
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
MES monohydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
Methanol, NMR reference standard
SAFC
MES monohydrate
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
MES hydrate
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%