Merck
  • Immediate utility of two approved agents to target both the metabolic mevalonate pathway and its restorative feedback loop.
Immediate utility of two approved agents to target both the metabolic mevalonate pathway and its restorative feedback loop.
Cancer research (2014-07-06)
Aleksandra Pandyra, Peter J Mullen, Manpreet Kalkat, Rosemary Yu, Janice T Pong, Zhihua Li, Suzanne Trudel, Karl S Lang, Mark D Minden, Aaron D Schimmer, Linda Z Penn
ABSTRACT
New therapies are urgently needed for hematologic malignancies, especially in patients with relapsed acute myelogenous leukemia (AML) and multiple myeloma. We and others have previously shown that FDA-approved statins, which are used to control hypercholesterolemia and target the mevalonate pathway (MVA), can trigger tumor-selective apoptosis. Our goal was to identify other FDA-approved drugs that synergize with statins to further enhance the anticancer activity of statins in vivo. Using a screen composed of other FDA approved drugs, we identified dipyridamole, used for the prevention of cerebral ischemia, as a potentiator of statin anticancer activity. The statin-dipyridamole combination was synergistic and induced apoptosis in multiple myeloma and AML cell lines and primary patient samples, whereas normal peripheral blood mononuclear cells were not affected. This novel combination also decreased tumor growth in vivo. Statins block HMG-CoA reductase (HMGCR), the rate-limiting enzyme of the MVA pathway. Dipyridamole blunted the feedback response, which upregulates HMGCR and HMG-CoA synthase 1 (HMGCS1) following statin treatment. We further show that dipyridamole inhibited the cleavage of the transcription factor required for this feedback regulation, sterol regulatory element-binding transcription factor 2 (SREBF2, SREBP2). Simultaneously targeting the MVA pathway and its restorative feedback loop is preclinically effective against hematologic malignancies. This work provides strong evidence for the immediate evaluation of this novel combination of FDA-approved drugs in clinical trials.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
SKU
Pack Size
Availability
Price
Quantity
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dimethyl sulfoxide, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water