Merck
  • Synthesis and biological evaluation of fluconazole analogs with triazole-modified scaffold as potent antifungal agents.
Synthesis and biological evaluation of fluconazole analogs with triazole-modified scaffold as potent antifungal agents.
Bioorganic & medicinal chemistry (2015-03-06)
Seyedeh Mahdieh Hashemi, Hamid Badali, Hamid Irannejad, Mohammad Shokrzadeh, Saeed Emami
ABSTRACT
In order to find new azole antifungals, we have recently designed a series of triazole alcohols in which one of the 1,2,4-triazol-1-yl group in fluconazole structure has been replaced with 4-amino-5-aryl-3-mercapto-1,2,4-triazole motif. In this paper, we focused on the structural refinement of the primary lead, by removing the amino group from the structure to achieve 5-aryl-3-mercapto-1,2,4-triazole derivatives 10a-i and 11a-i. The in vitro antifungal susceptibility testing of title compounds demonstrated that most compounds had potent inhibitory activity against Candida species. Among them, 5-(2,4-dichlorophenyl)triazole analogs 10h and 11h with MIC values of <0.01 to 0.5μg/mL were 4-256 times more potent than fluconazole against Candida species.
Product Description
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Fluconazole, European Pharmacopoeia (EP) Reference Standard