Merck
  • pH-sensitive chitosan/alginate core-shell nanoparticles for efficient and safe oral insulin delivery.
pH-sensitive chitosan/alginate core-shell nanoparticles for efficient and safe oral insulin delivery.
International journal of biological macromolecules (2014-09-23)
Piyasi Mukhopadhyay, Souma Chakraborty, Sourav Bhattacharya, Roshnara Mishra, P P Kundu
ABSTRACT
Chitosan-alginate (CS/ALG) nanoparticles were prepared by formation of an ionotropic pre-gelation of an alginate (ALG) core entrapping insulin, followed by chitosan (CS) polyelectrolyte complexation, for successful oral insulin administration. Mild preparation process without harsh chemicals is aimed at improving insulin bio-efficiency in in vivo model. The nanoparticles showed an average particle size of 100-200 nm in dynamic light scattering (DLS), with almost spherical or sub-spherical shape and ∼ 85% of insulin encapsulation. Again, retention of almost entire amount of encapsulated insulin in simulated gastric buffer followed by its sustained release in simulated intestinal condition proved its pH sensitivity in in vitro release studies. Significant hypoglycemic effects with improved insulin-relative bioavailability (∼ 8.11%) in in vivo model revealed the efficacy of these core-shell nanoparticles of CS/ALG as an oral insulin carrier. No systemic toxicity was found after its peroral treatment, suggesting these core-shell nanoparticles as a promising device for potential oral insulin delivery.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis