Merck
  • Simple and rapid high performance liquid chromatography method for the determination of polidocanol as bulk product and in pharmaceutical polymer matrices using charged aerosol detection.
Simple and rapid high performance liquid chromatography method for the determination of polidocanol as bulk product and in pharmaceutical polymer matrices using charged aerosol detection.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
David Ilko, Sebastian Puhl, Lorenz Meinel, Oliver Germershaus, Ulrike Holzgrabe
ABSTRACT
Currently, neither the European nor the United States Pharmacopoeia provide a method for the determination of polidocanol (PD) content despite the fact that PD, besides being an excipient, is also used as an active pharmaceutical ingredient. We therefore developed a method where the PD content was determined using a Kinetex C18 column operated at 40°C with water-acetonitrile (15:85, v/v) as mobile phase. A Corona(®) charged aerosol detector was employed for the detection of PD that is lacking a suitable UV chromophore. The method was fully validated. Additionally, the method was applied for the determination of PD release from a pharmaceutical polymer matrix consisting of poly-ɛ-caprolactone and poly(lactic-co-glycolic acid) and PD.
Product Description
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
1-Dodecanol, Selectophore, ≥98.0%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
1-Dodecanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)