Merck
  • Evaluation of new superficially porous particles with carbon core and nanodiamond-polymer shell for proteins characterization.
Evaluation of new superficially porous particles with carbon core and nanodiamond-polymer shell for proteins characterization.
Journal of pharmaceutical and biomedical analysis (2014-12-17)
Balázs Bobály, Davy Guillarme, Szabolcs Fekete
ABSTRACT
A new superficially porous material possessing a carbon core and nanodiamond-polymer shell and pore size of 180Å was evaluated for the analysis of large proteins. Because the stationary phase on this new support contains a certain amount of protonated amino groups within the shell structure, the resulting retention mechanism is most probably a mix between reversed phase and anion exchange. However, under the applied conditions (0.1-0.5% TFA in the mobile phase), it seemed that the main retention mechanism for proteins was hydrophobic interaction with the C18 alkylchains on this carbon based material. In this study, we demonstrated that there was no need to increase mobile phase temperature, as the peak capacity was not modified considerably between 30 and 80°C for model proteins. Thus, the risk of thermal on-column degradation or denaturation of large proteins is not relevant. Another important difference compared to silica-based materials is that this carbon-based column requires larger amount of TFA, comprised between 0.2 and 0.5%. Finally, it is important to mention that selectivity between closely related proteins (oxidized, native and reduced forms of Interferon α-2A variants) could be changed mostly through mobile phase temperature.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Methionine, European Pharmacopoeia (EP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%