Merck
  • Flaxseed lignan wound healing formulation: characterization and in vivo therapeutic evaluation.
Flaxseed lignan wound healing formulation: characterization and in vivo therapeutic evaluation.
International journal of biological macromolecules (2014-09-23)
D Draganescu, C Ibanescu, B I Tamba, C V Andritoiu, G Dodi, M I Popa
ABSTRACT
Flaxseed lignans are a natural source of useful biologically active components that show a diverse spectrum of health-promoting properties. The valuable effects of the phenolic molecules are mainly due to their antioxidant activity by preventing oxidative stress and stimulate collagen synthesis, therefore, providing benefits to the skin. The present work highlights the development of flaxseed extract formulation as novel wound healing agent. The recognition of key structural features within flaxseed extract was crucial for the design and development of the therapeutic cream. Chromatographic analyses were employed for bioactive compounds identification and quantification. Folin-Ciocalteu method determined the total phenolic content and the antioxidant properties were evaluated by DPPH assay. The storage and loss modulus and tan δ were calculated for cream rheological properties evaluation. In vitro diffusion capacity and in vivo wound healing activity of phenolic cream were evaluated on Wistar rats. The collective properties and healing effect of the flaxseed suggested wound healing capacity.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Toluene, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Hexane, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)