Merck
  • Chemometric assisted ultrasound leaching-solid phase extraction followed by dispersive-solidification liquid-liquid microextraction for determination of organophosphorus pesticides in soil samples.
Chemometric assisted ultrasound leaching-solid phase extraction followed by dispersive-solidification liquid-liquid microextraction for determination of organophosphorus pesticides in soil samples.
Talanta (2015-03-17)
Kamyar Ahmadi, Yaser Abdollahzadeh, Mehdi Asadollahzadeh, Alireza Hemmati, Hamed Tavakoli, Rezvan Torkaman
ABSTRACT
Ultrasound leaching-solid phase extraction (USL-SPE) followed by dispersive-solidification liquid-liquid microextraction (DSLLME) was developed for preconcentration and determination of organophosphorus pesticides (OPPs) in soil samples prior gas chromatography-mass spectrometry analysis. At first, OPPs were ultrasonically leached from soil samples by using methanol. After centrifugation, the separated methanol was diluted to 50 mL with double-distillated water and passed through the C18 SPE cartridge. OPPs were eluted with 1 mL acetonitrile. Thus, 1 mL acetonitrile extract (disperser solvent) and 10 µL 1-undecanol (extraction solvent) were added to 5 mL double-distilled water and a DSLLME technique was applied. The variables of interest in the USL-SPE-DSLLME method were optimized with the aid of chemometric approaches. First, in screening experiments, fractional factorial design (FFD) was used for selecting the variables which significantly affected the extraction procedure. Afterwards, the significant variables were optimized using response surface methodology (RSM) based on central composite design (CCD). Under the optimum conditions, the enrichment factors were 6890-8830. The linear range was 0.025-625 ng g(-1) and limits of detection (LODs) were between 0.012 and 0.2 ng g(-1). The relative standard deviations (RSDs) were in the range of 4.06-8.9% (n=6). The relative recoveries of OPPs from different soil samples were 85-98%.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfotep, PESTANAL®, analytical standard
Supelco
Acetone, analytical standard
Supelco
Methanol, analytical standard
Supelco
Hexadecane, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
1-Undecanol, 97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%