Merck
  • Proteomic characterization of human coronary thrombus in patients with ST-segment elevation acute myocardial infarction.
Proteomic characterization of human coronary thrombus in patients with ST-segment elevation acute myocardial infarction.
Journal of proteomics (2014-07-30)
Sergio Alonso-Orgaz, Rafael Moreno-Luna, Juan A López, Felix Gil-Dones, Luis R Padial, Jose Moreu, Fernando de la Cuesta, Maria G Barderas
ABSTRACT
Acute myocardial infarction with ST-segment elevation (STEMI) initiates with intraluminal thrombosis and results in total occlusion of the coronary artery. To date, characterization of the coronary thrombus proteome in STEMI patients has not been yet accomplished. Therefore, we aimed to perform an in-depth proteomic characterization of the human coronary thrombus by means of three different approaches: 2-DE followed by mass spectrometry (MALDI MS/MS), 1-DE combined either with liquid chromatography coupled to mass spectrometry in a MALDI TOF/TOF (LC-MALDI-MS/MS), or in a LTQ-Orbitrap (LC-ESI-MS/MS). This approach allowed us to identify a total of 708 proteins in the thrombus. Expression in coronary thrombi (n=20) of 14 proteins was verified, and the expression of fibrin and 6 cell markers (platelets, monocytes, neutrophils, eosinophils, T-cells and B-cells) quantified by selected reaction monitoring (SRM). A positive correlation of 5 proteins (fermitin homolog 3, thrombospondin-1, myosin-9, beta parvin and ras-related protein Rap-1b) with CD41 was found, pointing out the potential activation of a focal adhesion pathway within thrombus platelets. DIDO1 protein was found to correlate negatively with thrombus fibrin, and was found up-regulated in the plasma of these STEMI patients, which may constitute a starting point for further analyses in the search for biomarkers of thrombosis. The proteomic characterization of the human coronary thrombus may contribute to a better understanding of the mechanisms involved in acute coronary syndrome, and thus pave the road for the identification of new therapeutic targets that may help addressing this and other thrombotic diseases. A novel methodology to characterize thrombus composition and expression of a sub-group of proteins is hereby described, which allowed linking protein expression with cellular and ECM matrix composition of the thrombus. Five proteins (fermitin homolog 3, thrombospondin-1, myosin-9, beta parvin and ras-related protein Rap-1b) co-express within the human coronary thrombus with CD41, pointing out the potential activation of a focal adhesion pathway within thrombus platelets during thrombus formation. Besides, the protein death-inducer obliterator 1, found to be expressed within the human coronary thrombus, has been proved to increase in the plasma of STEMI patients, which constitutes an important starting point for further analyses in the search for biomarkers of thrombosis.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O