Merck
  • Controlled gene delivery can enhance therapeutic outcome for cancer immune therapy for melanoma.
Controlled gene delivery can enhance therapeutic outcome for cancer immune therapy for melanoma.
Current gene therapy (2014-11-22)
Shawna A Shirley, Cathryn G Lundberg, Fanying Li, Niculina Burcus, Richard Heller
ABSTRACT
Effective delivery still remains a major hurdle in the development of gene based therapies. While technological advances have occurred that have improved delivery in general, there is still a need for controlled delivery in order to achieve therapeutic effects. Gene electrotransfer (GET) can be utilized to accomplish this. Careful selection of parameters used for delivery such as amplitude, duration and number of pulses as well as plasmid construct can be manipulated in order to achieve appropriate levels of local expression. Previously we have shown that direct delivery of the therapeutic cytokine, interleukin 12 (IL-12), to tumors using electrotransfer can generate local and systemic anti-tumor effects in pre-clinical and clinical studies. Using this model we hypothesized that modulating local gene expression using GET can affect therapeutic outcome. To test this, we used multiple GET protocols and plasmids to achieve varying levels of local IL-12 expression. We found that high local gene expression did not give rise to a better therapeutic outcome. This suggests the level and possibly the duration of gene expression are important in mediating the host immune response against melanoma. These data also emphasize the importance of considering the desired immune outcome of the therapy when selecting parameters for GET.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology