Merck
  • Genetic and physiological responses of three freshwater diatoms to realistic diuron exposures.
Genetic and physiological responses of three freshwater diatoms to realistic diuron exposures.
Environmental science and pollution research international (2014-09-17)
S Moisset, S Kim Tiam, A Feurtet-Mazel, S Morin, F Delmas, N Mazzella, P Gonzalez
ABSTRACT
This study examined the effects of diuron on strains of three major freshwater diatom species, Eolimna minima, Nitzschia palea and Planothidium lanceolatum. These species are frequently recorded in the Morcille River, where diuron runs off during phytosanitary treatments of the vineyards around. Here, there were three diatom exposure groups for each species: 0, 1 and 10 μg/L diuron during a 14-day laboratory assessment. Diuron water concentration, cell number, photosynthetic activity and gene expression were assessed at 6 h and 2, 7 and 14 days after contamination. Diuron exposure altered photosynthetic activity in that the optimal quantum yield of photosystem II (PSII) decreased between 40 and 50% and, for P. lanceolatum at 10 μg/L, there was complete inhibition. Genetic responses indicated diuron effects on both photosystem II and mitochondrial metabolism in all three species at both diuron exposure levels. Thus, analysis of the expression of psaA, d1, cox1, nad5 and 12s could be an early biomarker to detect pesticide pollution. Overall, this study revealed differences in diuron sensitivity among the three species: E. minima and N. palea appeared to be more tolerant than P. lanceolatum. These results suggest that the development of molecular tools, and more precisely of biomarkers, will aid in early assessment of contamination and water quality.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)