Merck
  • Role of the vasohibin family in the regulation of fetoplacental vascularization and syncytiotrophoblast formation.
Role of the vasohibin family in the regulation of fetoplacental vascularization and syncytiotrophoblast formation.
PloS one (2014-09-04)
Kaori Suenaga, Shuji Kitahara, Yasuhiro Suzuki, Miho Kobayashi, Sachiko Horie, Junichi Sugawara, Nobuo Yaegashi, Yasufumi Sato
ABSTRACT
Vasohibin-1 (VASH1) and vasohibin-2 (VASH2), the 2 members of the vasohibin family, have been identified as novel regulators of angiogenesis. VASH1 ceases angiogenesis, whereas VASH2 stimulates sprouting. Here we characterized their functional role in the placenta. Immunohistochemical analysis of human placental tissue clarified their distinctive localization; VASH1 in endothelial cells and VASH2 in trophoblasts. We then used a mouse model to explore their function. Wild-type, Vash1((-/-)), and Vash2((-/-)) mice on a C57BL6 background were used in their first pregnancy. As expected, the fetal vascular area was increased in the Vash1((-/-)) mice, whereas it was decreased in the Vash2((-/-)) mice relative to wild-type. In addition, we noticed that the Vash2((-/-)) mice at 18.5dpc displayed thinner villi of the labyrinth and larger maternal lacunae. Careful observation by an electron microscopy revealed that the syncytiotrophoblast formation was defective in the Vash2((-/-)) mice. To test the possible involvement of VASH2 in the syncytiotrophoblast formation, we examined the fusion of BeWo cells, a human trophoblastoid choriocarcinoma cell line. The forskolin treatment induced the fusion of BeWo cells, and the knockdown of VASH2 expression significantly inhibited this cell fusion. Conversely, the overexpression of VASH2 by the infection with adenovirus vector encoding human VASH2 gene significantly increased the fusion of BeWo cells. Glial cell missing-1 and endogenous retrovirus envelope glycoprotein Syncytin 1 and Syncytin 2 are known to be involved in the fusion of trophoblasts. However, VASH2 did not alter their expression in BeWo cells. These results indicate that VASH1 and VASH2 showed distinctive localization and opposing function on the fetoplacental vascularization. Moreover, our study shows for the first time that VASH2 expressed in trophoblasts is involved in the regulation of cell fusion for syncytiotrophoblast formation.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting human VASH2 (1)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma